accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHNH3

CAS RN 51857-17-1

IUPAC Name: tert-butyl N-(6-azaniumylhexyl)carbamate
Synonyms: tert-butyl N-(6-ammoniohexyl)carbamate
Molecular Weight (g/mol): 217.33
Molecular Formula: C11H25N2O2
InChi Key: RVZPDKXEHIRFPM-UHFFFAOYSA-O
SMILES: CC(C)(C)OC(=O)NCCCCCC[NH3+]
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (6)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)