missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylmethylamine, 97%

Catalog No. p-7021424
Change view
Click to view available options
Quantity:
100 mL
250 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA13746AE 100 mL
AAA13746AK 250 mL
AAA13746AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA13746AE Supplier Thermo Scientific Chemicals Supplier No. A13746AE

CAS: 103-67-3 | C8H11N | 121.18 g/mol

N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.

Solubility
Soluble in water (65 g/L at 20°C).

Notes
Air Sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, carbon dioxide.

Chemical Identifiers

CAS 103-67-3
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008289
InChI Key RIWRFSMVIUAEBX-UHFFFAOYSA-N
Synonym n-methylbenzylamine, methylbenzylamine, n-benzylmethylamine, benzyl methyl amine, benzenemethanamine, n-methyl, benzylmethylamine, n-methy-n-benzylamine, n-benzyl-n-methylamine, benzylamine, n-methyl, n-methyl phenyl methanamine
PubChem CID 7669
IUPAC Name N-methyl-1-phenylmethanamine
SMILES CNCC1=CC=CC=C1

Specifications

Melting Point -55°C
Density 0.94
pH 10
Boiling Point 184°C to 188°C
Flash Point 77°C (170°F)
Odor Amine-like
Refractive Index 1.523
Linear Formula C6H5CH2NHCH3
Quantity 100 mL
UN Number UN2922
Beilstein 606221
Sensitivity Air Sensitive
Solubility Information Soluble in water (65g/L at 20°C).
Formula Weight 121.18
Percent Purity 97%
Chemical Name or Material N-Benzylmethylamine
Show More Show Less
Hazard Category H227-H301-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 203-133-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only