missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylidenebenzenesulfonamide 98.0+%, TCI America™

Catalog No. p-7119553
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22065G 5 g
B220625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22065G Supplier TCI America Supplier No. B22065G
Only null left

Chemical Identifiers

CAS 13909-34-7
Molecular Formula C13H11NO2S
Molecular Weight (g/mol) 245.30
MDL Number MFCD00011587
InChI Key MPNRJEPBAYEQBY-UHFFFAOYSA-N
PubChem CID 9602006
IUPAC Name N-(phenylmethylidene)benzenesulfonamide
SMILES O=S(=O)(N=CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 86°C
Quantity 5 g
Formula Weight 245.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzylidenebenzenesulfonamide