accessibility menu, dialog, popup

UserName

NSOO(E/Z)

CAS RN 13909-34-7

IUPAC Name: N-(phenylmethylidene)benzenesulfonamide
Synonyms: N-(phenylmethylidene)benzenesulfonamide
Molecular Weight (g/mol): 245.3
Molecular Formula: C13H11NO2S
InChi Key: MPNRJEPBAYEQBY-UHFFFAOYSA-N
SMILES: O=S(=O)(N=CC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results