missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylaminoacetaldehyde diethyl acetal, 96%

Catalog No. AAA1384106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1384106 5 g
AAA1384114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1384106 Supplier Thermo Scientific Chemicals Supplier No. A1384106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 61190-10-1 | C13H21NO2 | 223.316 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61190-10-1
Molecular Formula C13H21NO2
Molecular Weight (g/mol) 223.316
MDL Number MFCD00051599
InChI Key SXFVQTYQHWRYOS-UHFFFAOYSA-N
Synonym n-benzylaminoacetaldehyde diethyl acetal, benzyl 2,2-diethoxyethyl amine, n-benzyl-n-2,2-diethoxyethyl amine, 2,2-diethoxyethyl benzylamine, acmc-20ak6r, benzylaminoacetaldehyde diethylacetal, benzyl-2,2-diethoxy-ethyl-amine, benzylamino-acetaldehyde diethyl acetal, n-benzyl 2-aminoacetaldehyde diethyl acetal
PubChem CID 521961
IUPAC Name N-benzyl-2,2-diethoxyethanamine
SMILES CCOC(CNCC1=CC=CC=C1)OCC

Specifications

Boiling Point 115°C to 117°C (1 mmHg)
Refractive Index 1.489
Quantity 5 g
Beilstein 2694610
Formula Weight 223.32
Percent Purity 96%
Chemical Name or Material N-Benzylaminoacetaldehyde diethyl acetal
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only