accessibility menu, dialog, popup

UserName

CAS RN 61190-10-1

IUPAC Name: benzyl(2,2-diethoxyethyl)azanium
Synonyms: benzyl(2,2-diethoxyethyl)azanium
Molecular Weight (g/mol): 224.32
Molecular Formula: C13H22NO2
InChi Key: SXFVQTYQHWRYOS-UHFFFAOYSA-O
SMILES: CCOC(C[NH2+]CC1=CC=CC=C1)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results