missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-3-pyridinemethylamine, 97%, Thermo Scientific Chemicals

Catalog No. AAH50978MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50978MD 250 mg
AAH5097803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50978MD Supplier Thermo Scientific Chemicals Supplier No. H50978MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 63361-56-8 | Molecular Formula: C13H14N2 | Molecular Weight: 198.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63361-56-8
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.27
MDL Number MFCD00716866,MFCD08061098
InChI Key COHYOBKZKMKMIX-UHFFFAOYSA-N
Synonym benzyl-pyridin-3-ylmethyl-amine, n-nicotinylbenzylamine, benzyl pyridin-3-ylmethyl amine, benzylpyridin-3-ylmethylamine, 1-phenyl-n-pyridin-3-ylmethyl methylamine, n-benzyl-3-pyridinemethylamine, n-benzyl-1-pyridin-3-yl methanamine, benzyl 3-pyridylmethyl amine, acmc-20aos7, 3-benzylaminomethyl pyridine
PubChem CID 293794
IUPAC Name 1-phenyl-N-(pyridin-3-ylmethyl)methanamine
SMILES C(NCC1=CC=CN=C1)C1=CC=CC=C1

Specifications

Quantity 250 mg
Formula Weight 198.27
Percent Purity 97%
Chemical Name or Material N-Benzyl-3-pyridinemethylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only