CAS RN 63361-56-8
IUPAC Name:
benzyl[(pyridin-3-yl)methyl]amine
Synonyms:
benzyl(pyridin-3-ylmethyl)amine
Molecular Weight (g/mol):
198.27
Molecular Formula:
C13H14N2
InChi Key:
COHYOBKZKMKMIX-UHFFFAOYSA-N
SMILES:
C(NCC1=CC=CN=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
N-Benzyl-3-pyridinemethylamine, 97%, Thermo Scientific Chemicals
N-Benzyl-3-pyridinemethylamine, 97%, Thermo Scientific Chemicals
CAS: 63361-56-8 Molecular Formula: C13H14N2 Molecular Weight (g/mol): 198.27 MDL Number: MFCD00716866,MFCD08061098 InChI Key: COHYOBKZKMKMIX-UHFFFAOYSA-N Synonym: benzyl-pyridin-3-ylmethyl-amine,n-nicotinylbenzylamine,benzyl pyridin-3-ylmethyl amine,benzylpyridin-3-ylmethylamine,1-phenyl-n-pyridin-3-ylmethyl methylamine,n-benzyl-3-pyridinemethylamine,n-benzyl-1-pyridin-3-yl methanamine,benzyl 3-pyridylmethyl amine,acmc-20aos7,3-benzylaminomethyl pyridine PubChem CID: 293794 IUPAC Name: 1-phenyl-N-(pyridin-3-ylmethyl)methanamine SMILES: C(NCC1=CC=CN=C1)C1=CC=CC=C1