missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-3,3'-iminodipropionic Acid 98.0+%, TCI America™

Catalog No. B38265G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B38265G Supplier TCI America Supplier No. B38265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6405-28-3
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD16327542
InChI Key KSTIKMRBVZTNLX-UHFFFAOYSA-N
PubChem CID 56973687
IUPAC Name 3-[benzyl(2-carboxyethyl)amino]propanoic acid
SMILES OC(=O)CCN(CCC(O)=O)CC1=CC=CC=C1

Specifications

Melting Point 159°C
Color White
Quantity 5 g
Formula Weight 251.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzyl-3,3′-iminodipropionic Acid