accessibility menu, dialog, popup

UserName

NOHOOOH

CAS RN 6405-28-3

IUPAC Name: 3-[benzyl(2-carboxyethyl)amino]propanoic acid
Synonyms: 3-[benzyl(2-carboxyethyl)amino]propanoic acid
Molecular Weight (g/mol): 251.28
Molecular Formula: C13H17NO4
InChi Key: KSTIKMRBVZTNLX-UHFFFAOYSA-N
SMILES: OC(=O)CCN(CCC(O)=O)CC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results