missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-1,3-propanediamine 96.0+%, TCI America™

Catalog No. B49375ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49375ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B49375ML Supplier TCI America Supplier No. B49375ML
Only null left

Chemical Identifiers

CAS 13910-48-0
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.25
MDL Number MFCD00274361
InChI Key RFLHDXQRFPJPRR-UHFFFAOYSA-N
Synonym n1-benzylpropane-1,3-diamine, 3-benzylamino propylamine, n-benzylpropane-1,3-diamine, 3-aminopropyl benzyl amine, n-benzyl-1,3-propanediamine, n-3-aminopropyl-n-benzylamine, 3-aminopropyl benzylamine, n-benzylpropan-1,3-diamin, n-benzyltrimethylenediamine, n-3-aminopropyl benzylamine
PubChem CID 83811
IUPAC Name (3-aminopropyl)(benzyl)amine
SMILES NCCCNCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 102°C
Quantity 5 mL
UN Number 2735
Formula Weight 164.25
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Benzyl-1,3-propanediamine