accessibility menu, dialog, popup

UserName

H2NNH

CAS RN 13910-48-0

IUPAC Name: (3-aminopropyl)(benzyl)amine
Synonyms: (3-aminopropyl)(benzyl)amine
Molecular Weight (g/mol): 164.25
Molecular Formula: C10H16N2
InChi Key: RFLHDXQRFPJPRR-UHFFFAOYSA-N
SMILES: NCCCNCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results