missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyladenosine, Thermo Scientific Chemicals

Catalog No. AC464690050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464690050 5 g
AC464690010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464690050 Supplier Thermo Scientific Chemicals Supplier No. 464690050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4546-55-8 | C17H17N5O5 | 371.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4546-55-8
Molecular Formula C17H17N5O5
Molecular Weight (g/mol) 371.35
MDL Number MFCD00056988
InChI Key NZDWTKFDAUOODA-UCQOUBKUNA-N
IUPAC Name N-{9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-9H-purin-6-yl}benzamide
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12

Specifications

Melting Point 141.0°C to 145.0°C
Quantity 5 g
Formula Weight 371.35
Chemical Name or Material N-Benzoyladenosine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

RUO – Research Use Only