missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-D-glucosamine 98.0+%, TCI America™

Catalog No. p-7118372
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B02005G 5 g
B020025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B02005G Supplier TCI America Supplier No. B02005G
Only null left

Chemical Identifiers

CAS 655-42-5
Molecular Formula C13H17NO6
Molecular Weight (g/mol) 283.28
MDL Number MFCD00070521
InChI Key QCYHKZRHOGVACA-IRCOFANPSA-N
Synonym 2-Benzamido-2-deoxy-D-glucopyranose
PubChem CID 21117441
IUPAC Name N-[(2R,3R,4S,5R)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl]benzamide
SMILES C1=CC=C(C=C1)C(=O)NC(C=O)C(C(C(CO)O)O)O

Specifications

Color White
Quantity 5 g
Formula Weight 283.28
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzoyl-D-glucosamine