accessibility menu, dialog, popup

UserName

CAS RN 655-42-5

HOOHOHOHNHOO

CAS RN 655-42-5

IUPAC Name: N-(3,4,5,6-tetrahydroxy-1-oxohexan-2-yl)benzamide
Synonyms: N-(3,4,5,6-tetrahydroxy-1-oxohexan-2-yl)benzamide
Molecular Weight (g/mol): 283.28
Molecular Formula: C13H17NO6
InChi Key: QCYHKZRHOGVACA-UHFFFAOYNA-N
SMILES: OCC(O)C(O)C(O)C(NC(=O)C1=CC=CC=C1)C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results