missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Allyl-p-toluenesulfonamide 98.0+%, TCI America™
$197.59 - $672.92
Chemical Identifiers
| CAS | 50487-71-3 |
|---|---|
| Molecular Formula | C10H13NO2S |
| Molecular Weight (g/mol) | 211.279 |
| MDL Number | MFCD00182633 |
| InChI Key | SQMCYQSCMCMEIL-UHFFFAOYSA-N |
| PubChem CID | 297877 |
| IUPAC Name | 4-methyl-N-prop-2-enylbenzenesulfonamide |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)NCC=C |
Chemical Identifiers
| 50487-71-3 | |
| 211.279 | |
| SQMCYQSCMCMEIL-UHFFFAOYSA-N | |
| 4-methyl-N-prop-2-enylbenzenesulfonamide |
| C10H13NO2S | |
| MFCD00182633 | |
| 297877 | |
| CC1=CC=C(C=C1)S(=O)(=O)NCC=C |
Specifications
| 50487-71-3 | |
| White | |
| MFCD00182633 | |
| SQMCYQSCMCMEIL-UHFFFAOYSA-N | |
| 4-methyl-N-prop-2-enylbenzenesulfonamide | |
| 297877 | |
| ≥98.0% (GC) | |
| N-Allyl-p-toluenesulfonamide |
| 63°C | |
| C10H13NO2S | |
| 1 g | |
| CC1=CC=C(C=C1)S(=O)(=O)NCC=C | |
| 211.279 | |
| 211.28 | |
| Crystalline Powder |
Safety and Handling
TSCA : No