accessibility menu, dialog, popup

UserName

CH3SOOHNCH2

CAS RN 50487-71-3

IUPAC Name: 4-methyl-N-(prop-2-en-1-yl)benzene-1-sulfonamide
Synonyms: 4-methyl-N-(prop-2-en-1-yl)benzenesulfonamide
Molecular Weight (g/mol): 211.28
Molecular Formula: C10H13NO2S
InChi Key: SQMCYQSCMCMEIL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NCC=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results