missing translation for 'onlineSavingsMsg'
Learn More

N-Allyl-p-toluenesulfonamide 98.0+%, TCI America™

Catalog No. A26651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26651G 1 g
A26655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26651G Supplier TCI America Supplier No. A26651G
Only null left

Chemical Identifiers

CAS 50487-71-3
Molecular Formula C10H13NO2S
Molecular Weight (g/mol) 211.279
MDL Number MFCD00182633
InChI Key SQMCYQSCMCMEIL-UHFFFAOYSA-N
PubChem CID 297877
IUPAC Name 4-methyl-N-prop-2-enylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NCC=C

Specifications

Melting Point 63°C
Color White
Quantity 1 g
Formula Weight 211.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Allyl-p-toluenesulfonamide