missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-o-phenetidine 98.0+%, TCI America™

Catalog No. A0103500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0103500G 500 g
A010325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0103500G Supplier TCI America Supplier No. A0103500G
Only null left

Chemical Identifiers

CAS 581-08-8
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00059228
InChI Key SQRTWLGWCOJOTO-UHFFFAOYSA-N
PubChem CID 11383
IUPAC Name N-(2-ethoxyphenyl)acetamide
SMILES CCOC1=CC=CC=C1NC(=O)C

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 500 g
Synonym N-Acetyl-2-ethoxyaniline, 2′-Ethoxyacetanilide
Formula Weight 179.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-o-phenetidine
EINECSNumber (3)-0695
RTECSNumber AM4350000
TSCA Yes