accessibility menu, dialog, popup

UserName

CH3OHNH3CO

CAS RN 581-08-8

IUPAC Name: N-(2-ethoxyphenyl)acetamide
Synonyms: acetylphenetidin
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: SQRTWLGWCOJOTO-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1NC(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results