Learn More
N-Acetyl-N'-phenylhydrazine, 98%
CAS: 114-83-0 | C8H10N2O | 150.181 g/mol
$187.12 - $663.75
Chemical Identifiers
| CAS | 114-83-0 |
|---|---|
| Molecular Formula | C8H10N2O |
| Molecular Weight (g/mol) | 150.181 |
| MDL Number | MFCD00008672 |
| InChI Key | UICBCXONCUFSOI-UHFFFAOYSA-N |
| Synonym | 1-acetyl-2-phenylhydrazine, hydracetin, acetylphenylhydrazine, pyrodin, pyrodine, acetic acid, 2-phenylhydrazide, n-acetyl-n'-phenylhydrazine, n'-phenylacethydrazide, acetic acid phenylhydrazone, n 1-acetylphenylhydrazine |
| PubChem CID | 8247 |
| IUPAC Name | N'-phenylacetohydrazide |
| SMILES | CC(=O)NNC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1444622
|
Thermo Scientific Chemicals
A1444622 |
100 g |
N/A
|
|
|||||
|
AAA1444636
|
Thermo Scientific Chemicals
A1444636 |
500 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 114-83-0 | |
| 150.181 | |
| UICBCXONCUFSOI-UHFFFAOYSA-N | |
| 8247 | |
| CC(=O)NNC1=CC=CC=C1 |
| C8H10N2O | |
| MFCD00008672 | |
| 1-acetyl-2-phenylhydrazine, hydracetin, acetylphenylhydrazine, pyrodin, pyrodine, acetic acid, 2-phenylhydrazide, n-acetyl-n'-phenylhydrazine, n'-phenylacethydrazide, acetic acid phenylhydrazone, n 1-acetylphenylhydrazine | |
| N'-phenylacetohydrazide |
Specifications
| 114-83-0 | |
| C8H10N2O | |
| 100 g | |
| 742880 | |
| UICBCXONCUFSOI-UHFFFAOYSA-N | |
| N'-phenylacetohydrazide | |
| 8247 | |
| 98% |
| 128°C to 131°C | |
| MFCD00008672 | |
| UN2811 | |
| 1-acetyl-2-phenylhydrazine, hydracetin, acetylphenylhydrazine, pyrodin, pyrodine, acetic acid, 2-phenylhydrazide, n-acetyl-n'-phenylhydrazine, n'-phenylacethydrazide, acetic acid phenylhydrazone, n 1-acetylphenylhydrazine | |
| CC(=O)NNC1=CC=CC=C1 | |
| 150.181 | |
| 150.18 | |
| N-Acetyl-N'-phenylhydrazine |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 204-055-3
RTECSNumber : AJ2900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only