missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-N'-phenylhydrazine, 98%

Catalog No. AAA1444622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1444622 100 g
AAA1444636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1444622 Supplier Thermo Scientific Chemicals Supplier No. A1444622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 114-83-0 | C8H10N2O | 150.181 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 114-83-0
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00008672
InChI Key UICBCXONCUFSOI-UHFFFAOYSA-N
Synonym 1-acetyl-2-phenylhydrazine, hydracetin, acetylphenylhydrazine, pyrodin, pyrodine, acetic acid, 2-phenylhydrazide, n-acetyl-n'-phenylhydrazine, n'-phenylacethydrazide, acetic acid phenylhydrazone, n 1-acetylphenylhydrazine
PubChem CID 8247
IUPAC Name N'-phenylacetohydrazide
SMILES CC(=O)NNC1=CC=CC=C1

Specifications

Melting Point 128°C to 131°C
Quantity 100 g
UN Number UN2811
Beilstein 742880
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material N-Acetyl-N'-phenylhydrazine
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 204-055-3
RTECSNumber AJ2900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only