missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-DL-tryptophan, 98+%, extra pure

CAS: 87-32-1 | C13H14N2O3 | 246.27 g/mol

Supplier:  Thermo Scientific Chemicals 227151000

 View more versions of this product

Catalog No. AC227151000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Acetyl-DL-tryptophan
87-32-1
100.0
White to Yellow
Authentic
C13H14N2O3
100 g
Solubility in water: 0,51g/L (20°C). Other solubilities: soluble in methanol
CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
246.27
CHEBI:70976
98+%
Fine Crystalline Powder
99+%
99.0
204.0°C to 206.0°C
0.5% max. (105°C)
99+%
MFCD00005644
n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl
DZTHIGRZJZPRDV-UHFFFAOYSA-N
2-acetamido-3-(1H-indol-3-yl)propanoic acid
2002
246.27
Extra Pure

Chemical Identifiers

87-32-1
246.27
DZTHIGRZJZPRDV-UHFFFAOYSA-N
2002
2-acetamido-3-(1H-indol-3-yl)propanoic acid
C13H14N2O3
MFCD00005644
n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl
CHEBI:70976
CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-739-3

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only