CAS RN 87-32-1
Thermo Scientific Chemicals N-Acetyl-DL-tryptophan, 98+%, extra pure
CAS: 87-32-1 Molecular Formula: C13H14N2O3 Molecular Weight (g/mol): 246.27 MDL Number: MFCD00005644 InChI Key: DZTHIGRZJZPRDV-UHFFFAOYSA-N Synonym: n-acetyl-dl-tryptophan,n-acetyltryptophan,ac-dl-trp-oh,acetyltryptophan,dl-acetyltryptophan,2-acetamido-3-1h-indol-3-yl propanoic acid,dl-n-acetyltryptophan,n-acetyl-dl-tryptophane,l-tryptophan, n-acetyl,dl-tryptophan, n-acetyl PubChem CID: 2002 ChEBI: CHEBI:70976 IUPAC Name: 2-acetamido-3-(1H-indol-3-yl)propanoic acid SMILES: CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
N-Acetyl-DL-tryptophan, 99%
CAS: 87-32-1 Molecular Formula: C13H14N2O3 Molecular Weight (g/mol): 246.27 MDL Number: MFCD00005644 InChI Key: DZTHIGRZJZPRDV-UHFFFAOYSA-N Synonym: n-acetyl-dl-tryptophan,n-acetyltryptophan,ac-dl-trp-oh,acetyltryptophan,dl-acetyltryptophan,2-acetamido-3-1h-indol-3-yl propanoic acid,dl-n-acetyltryptophan,n-acetyl-dl-tryptophane,l-tryptophan, n-acetyl,dl-tryptophan, n-acetyl PubChem CID: 2002 ChEBI: CHEBI:70976 IUPAC Name: 2-acetamido-3-(1H-indol-3-yl)propanoic acid SMILES: CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
N-Acetyl-DL-tryptophan 98.0+%, TCI America™
CAS: 87-32-1 Molecular Formula: C13H14N2O3 Molecular Weight (g/mol): 246.27 MDL Number: MFCD00005644 InChI Key: DZTHIGRZJZPRDV-UHFFFAOYSA-N Synonym: n-acetyl-dl-tryptophan,n-acetyltryptophan,ac-dl-trp-oh,acetyltryptophan,dl-acetyltryptophan,2-acetamido-3-1h-indol-3-yl propanoic acid,dl-n-acetyltryptophan,n-acetyl-dl-tryptophane,l-tryptophan, n-acetyl,dl-tryptophan, n-acetyl PubChem CID: 2002 ChEBI: CHEBI:70976 IUPAC Name: 2-acetamido-3-(1H-indol-3-yl)propanoic acid SMILES: CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
N-acetyl-DL-tryptophan, ∽99%, MP Biomedicals™
CAS: 87-32-1 Molecular Formula: C13H14N2O3 Molecular Weight (g/mol): 246.27 MDL Number: MFCD00005644 InChI Key: DZTHIGRZJZPRDV-UHFFFAOYSA-N Synonym: n-acetyl-dl-tryptophan,n-acetyltryptophan,ac-dl-trp-oh,acetyltryptophan,dl-acetyltryptophan,2-acetamido-3-1h-indol-3-yl propanoic acid,dl-n-acetyltryptophan,n-acetyl-dl-tryptophane,l-tryptophan, n-acetyl,dl-tryptophan, n-acetyl PubChem CID: 2002 ChEBI: CHEBI:70976 IUPAC Name: 2-acetamido-3-(1H-indol-3-yl)propanoic acid SMILES: CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O