missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-DL-tryptophan, 98+%, extra pure

Catalog No. p-50009
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
Catalog No. Quantity
AC227150250 25 g
AC227151000 100 g
2 options

Catalog No. AC227150250

Supplier: Thermo Scientific Chemicals 227150250

Only null left

CAS: 87-32-1 | C13H14N2O3 | 246.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-32-1
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.27
MDL Number MFCD00005644
InChI Key DZTHIGRZJZPRDV-UHFFFAOYSA-N
Synonym n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl
PubChem CID 2002
ChEBI CHEBI:70976
IUPAC Name 2-acetamido-3-(1H-indol-3-yl)propanoic acid
SMILES CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 204.0°C to 206.0°C
Color White to Yellow
Loss on Drying 0.5% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: 0,51g/L (20°C). Other solubilities: soluble in methanol
Formula Weight 246.27
Percent Purity 98+%
Grade Extra Pure
Physical Form Fine Crystalline Powder
Chemical Name or Material N-Acetyl-DL-tryptophan, 99+%
Show More Show Less
EINECSNumber 201-739-3
TSCA TSCA

RUO – Research Use Only