missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N-Acetyl-DL-tryptophan, 98+%, extra pure
CAS: 87-32-1 | C13H14N2O3 | 246.27 g/mol
Supplier: Thermo Scientific Chemicals 227150250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-Acetyl-DL-tryptophan | |
| 87-32-1 | |
| 100.0 | |
| White to Yellow | |
| Authentic | |
| Glass bottle | |
| MFCD00005644 | |
| n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl | |
| DZTHIGRZJZPRDV-UHFFFAOYSA-N | |
| 2-acetamido-3-(1H-indol-3-yl)propanoic acid | |
| 2002 | |
| 246.27 | |
| Extra Pure |
| 99+% | |
| 99.0 | |
| 204.0°C to 206.0°C | |
| 0.5% max. (105°C) | |
| 98+% | |
| C13H14N2O3 | |
| 25 g | |
| Solubility in water: 0,51g/L (20°C). Other solubilities: soluble in methanol | |
| CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O | |
| 246.27 | |
| CHEBI:70976 | |
| 98+% | |
| Fine Crystalline Powder |
Chemical Identifiers
| 87-32-1 | |
| 246.27 | |
| DZTHIGRZJZPRDV-UHFFFAOYSA-N | |
| 2002 | |
| 2-acetamido-3-(1H-indol-3-yl)propanoic acid |
| C13H14N2O3 | |
| MFCD00005644 | |
| n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl | |
| CHEBI:70976 | |
| CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O |
Safety and Handling
EINECSNumber : 201-739-3
TSCA : TSCA
RUO – Research Use Only