missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-DL-2-phenylglycine 98.0+%, TCI America™
Supplier: TCI America A268725G
Specifications
| N-Acetyl-DL-2-phenylglycine | |
| 198°C | |
| C10H11NO3 | |
| 25 g | |
| VKDFZMMOLPIWQQ-UHFFFAOYNA-N | |
| 2-acetamido-2-phenylacetic acid | |
| 85935 | |
| ≥98.0% (HPLC,T) |
| 15962-46-6 | |
| White-Yellow | |
| MFCD00055924 | |
| acetylamino phenyl acetic acid, ac-dl-phg-oh, 2-acetylamino-2-phenylacetic acid, acetamido phenyl acetic acid, n-acetyl-l-phenylglycine, n-acetyl-dl-2-phenylglycine, 1-acetamido phenylacetic acid, phenylaceturic acid, n-acetyl-2-phenylglycine, maybridge1_002435 | |
| CC(=O)NC(C(O)=O)C1=CC=CC=C1 | |
| 193.20 | |
| 193.20 | |
| Crystalline Powder |
Chemical Identifiers
| 15962-46-6 | |
| 193.20 | |
| VKDFZMMOLPIWQQ-UHFFFAOYNA-N | |
| 85935 | |
| CC(=O)NC(C(O)=O)C1=CC=CC=C1 |
| C10H11NO3 | |
| MFCD00055924 | |
| acetylamino phenyl acetic acid, ac-dl-phg-oh, 2-acetylamino-2-phenylacetic acid, acetamido phenyl acetic acid, n-acetyl-l-phenylglycine, n-acetyl-dl-2-phenylglycine, 1-acetamido phenylacetic acid, phenylaceturic acid, n-acetyl-2-phenylglycine, maybridge1_002435 | |
| 2-acetamido-2-phenylacetic acid |
Safety and Handling
EINECSNumber : (9)-1623
TSCA : No