missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-DL-2-phenylglycine 98.0+%, TCI America™

Catalog No. p-7118023
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26875G 5 g
A268725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26875G Supplier TCI America Supplier No. A26875G

Chemical Identifiers

CAS 15962-46-6
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00055924
InChI Key VKDFZMMOLPIWQQ-UHFFFAOYNA-N
Synonym acetylamino phenyl acetic acid, ac-dl-phg-oh, 2-acetylamino-2-phenylacetic acid, acetamido phenyl acetic acid, n-acetyl-l-phenylglycine, n-acetyl-dl-2-phenylglycine, 1-acetamido phenylacetic acid, phenylaceturic acid, n-acetyl-2-phenylglycine, maybridge1_002435
PubChem CID 85935
IUPAC Name 2-acetamido-2-phenylacetic acid
SMILES CC(=O)NC(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 198°C
Color White-Yellow
Quantity 5 g
Formula Weight 193.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-DL-2-phenylglycine
EINECSNumber (9)-1623
TSCA No