CAS RN 15962-46-6
IUPAC Name:
2-acetamido-2-phenylacetic acid
Synonyms:
acetamido(phenyl)acetic acid
Molecular Weight (g/mol):
193.2
Molecular Formula:
C10H11NO3
InChi Key:
VKDFZMMOLPIWQQ-UHFFFAOYNA-N
SMILES:
CC(=O)NC(C(O)=O)C1=CC=CC=C1
1
–
1
of
1
results
1
N-Acetyl-DL-2-phenylglycine 98.0+%, TCI America™
CAS: 15962-46-6 Molecular Formula: C10H11NO3 Molecular Weight (g/mol): 193.20 MDL Number: MFCD00055924 InChI Key: VKDFZMMOLPIWQQ-UHFFFAOYNA-N Synonym: acetylamino phenyl acetic acid,ac-dl-phg-oh,2-acetylamino-2-phenylacetic acid,acetamido phenyl acetic acid,n-acetyl-l-phenylglycine,n-acetyl-dl-2-phenylglycine,1-acetamido phenylacetic acid,phenylaceturic acid,n-acetyl-2-phenylglycine,maybridge1_002435 PubChem CID: 85935 IUPAC Name: 2-acetamido-2-phenylacetic acid SMILES: CC(=O)NC(C(O)=O)C1=CC=CC=C1