missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-norvaline 98.0+%, TCI America™
$130.48 - $426.87
Chemical Identifiers
| CAS | 252049-05-1 |
|---|---|
| Molecular Formula | C21H23NO4 |
| Molecular Weight (g/mol) | 353.42 |
| MDL Number | MFCD01861326 |
| InChI Key | HKELUUGCKFRJQM-IBGZPJMESA-N |
| Synonym | fmoc-n-methyl-l-norvaline, fmoc-n-me-nva-oh, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino pentanoic acid, fmoc-n-me-l-2-aminovaleric acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino pentanoic acid, l-norvaline, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, l-norvaline,n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, fmoc-n-me-l-norvaline, n-fmoc-n-methyl-l-norvaline, fmoc-nalpha-methyl-l-norvaline |
| PubChem CID | 7009909 |
| IUPAC Name | (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)pentanoic acid |
| SMILES | CCC[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Chemical Identifiers
| 252049-05-1 | |
| 353.42 | |
| HKELUUGCKFRJQM-IBGZPJMESA-N | |
| 7009909 | |
| CCC[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| C21H23NO4 | |
| MFCD01861326 | |
| fmoc-n-methyl-l-norvaline, fmoc-n-me-nva-oh, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino pentanoic acid, fmoc-n-me-l-2-aminovaleric acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino pentanoic acid, l-norvaline, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, l-norvaline,n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, fmoc-n-me-l-norvaline, n-fmoc-n-methyl-l-norvaline, fmoc-nalpha-methyl-l-norvaline | |
| (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)pentanoic acid |
Specifications
| 252049-05-1 | |
| White | |
| MFCD01861326 | |
| fmoc-n-methyl-l-norvaline, fmoc-n-me-nva-oh, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino pentanoic acid, fmoc-n-me-l-2-aminovaleric acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino pentanoic acid, l-norvaline, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, l-norvaline,n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, fmoc-n-me-l-norvaline, n-fmoc-n-methyl-l-norvaline, fmoc-nalpha-methyl-l-norvaline | |
| CCC[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
| 353.42 | |
| 353.42 | |
| Crystalline Powder |
| 132°C | |
| C21H23NO4 | |
| 1 g | |
| HKELUUGCKFRJQM-IBGZPJMESA-N | |
| (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)pentanoic acid | |
| 7009909 | |
| ≥98.0% (HPLC,T) | |
| N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-norvaline |
Safety and Handling
TSCA : No