accessibility menu, dialog, popup

UserName

CAS RN 252049-05-1

H3C(S)OHONCH3OO

CAS RN 252049-05-1

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)pentanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}pentanoic acid
Molecular Weight (g/mol): 353.42
Molecular Formula: C21H23NO4
InChi Key: HKELUUGCKFRJQM-IBGZPJMESA-N
SMILES: CCC[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results