missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine Monohydrate 98.0+%, TCI America™
$29.83 - $147.25
Chemical Identifiers
| CAS | 73731-37-0 |
|---|---|
| Molecular Formula | C19H21NO6 |
| Molecular Weight (g/mol) | 359.38 |
| MDL Number | MFCD00792901 |
| InChI Key | CGNQPFAECJFQNV-NRNQBQMASA-N |
| Synonym | fmoc-thr-oh, fmoc-l-threonine, fmoc-l-thr-oh, 2-9h-fluoren-9-ylmethoxycarbonylamino-3-hydroxy-butanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-threonine, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-hydroxybutanoic acid, n-9-fluorenylmethoxycarbonyl-l-threonine, 2s,3r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxybutanoic acid, fmoc-thr-oh monohydrate, n-9h-fluoren-9-ylmethoxy carbonyl-l-threonine monohydrate |
| PubChem CID | 6992530 |
| IUPAC Name | (2S,3R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxybutanoic acid hydrate |
| SMILES | O.C[C@@H](O)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O |
Chemical Identifiers
| 73731-37-0 | |
| 359.38 | |
| CGNQPFAECJFQNV-NRNQBQMASA-N | |
| 6992530 | |
| O.C[C@@H](O)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O |
| C19H21NO6 | |
| MFCD00792901 | |
| fmoc-thr-oh, fmoc-l-threonine, fmoc-l-thr-oh, 2-9h-fluoren-9-ylmethoxycarbonylamino-3-hydroxy-butanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-threonine, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-hydroxybutanoic acid, n-9-fluorenylmethoxycarbonyl-l-threonine, 2s,3r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxybutanoic acid, fmoc-thr-oh monohydrate, n-9h-fluoren-9-ylmethoxy carbonyl-l-threonine monohydrate | |
| (2S,3R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxybutanoic acid hydrate |
Specifications
| 73731-37-0 | |
| C19H21NO6 | |
| 1 g | |
| CGNQPFAECJFQNV-NRNQBQMASA-N | |
| (2S,3R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxybutanoic acid hydrate | |
| 6992530 | |
| ≥98.0% (HPLC,T) | |
| N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine Monohydrate |
| White-Yellow | |
| MFCD00792901 | |
| fmoc-thr-oh, fmoc-l-threonine, fmoc-l-thr-oh, 2-9h-fluoren-9-ylmethoxycarbonylamino-3-hydroxy-butanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-threonine, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-hydroxybutanoic acid, n-9-fluorenylmethoxycarbonyl-l-threonine, 2s,3r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxybutanoic acid, fmoc-thr-oh monohydrate, n-9h-fluoren-9-ylmethoxy carbonyl-l-threonine monohydrate | |
| O.C[C@@H](O)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O | |
| 359.38 | |
| 341.36 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer