missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-propargylglycine 98.0+%, TCI America™

Catalog No. F09261G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F09261G Supplier TCI America Supplier No. F09261G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 198561-07-8
Molecular Formula C20H17NO4
Molecular Weight (g/mol) 335.359
MDL Number MFCD01075095
InChI Key DJGMNCKHNMRKFM-SFHVURJKSA-N
Synonym fmoc-l-propargylglycine, fmoc-pra-oh, fmoc-propargyl-gly-oh, s-2-fmoc-amino-4-pentynoic acid, s-n-fmoc-2-2'-propynyl glycine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-ynoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-ynoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino pent-4-ynoic acid, 4-pentynoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s, 4-pentynoicacid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s
PubChem CID 2734461
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-ynoic acid
SMILES C#CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 176°C
Color White
Quantity 1 g
Formula Weight 335.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-propargylglycine
TSCA No
Recommended Storage Refrigerator