missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-leucine 98.0+%, TCI America™
Supplier: TCI America F029525G
Specifications
| N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-leucine | |
| 154°C | |
| C21H23NO4 | |
| 25 g | |
| CBPJQFCAFFNICX-TWYLJJHKNA-N | |
| (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methylpentanoic acid | |
| 1549133 | |
| ≥98.0% (HPLC,T) |
| 35661-60-0 | |
| White | |
| MFCD00037133 | |
| fmoc-l-leucine, fmoc-leu-oh, fmoc-l-leu-oh, n-fmoc-l-leucine, n-9-fluorenylmethoxycarbonyl-l-leucine, fmoc-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-l-leucine, unii-kf18f70uf3, fmoc-leu, n-alpha-fmoc-l-leucine | |
| CC(C)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
| 353.42 | |
| 353.42 | |
| Crystalline Powder |
Chemical Identifiers
| 35661-60-0 | |
| 353.42 | |
| CBPJQFCAFFNICX-TWYLJJHKNA-N | |
| 1549133 | |
| CC(C)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| C21H23NO4 | |
| MFCD00037133 | |
| fmoc-l-leucine, fmoc-leu-oh, fmoc-l-leu-oh, n-fmoc-l-leucine, n-9-fluorenylmethoxycarbonyl-l-leucine, fmoc-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-l-leucine, unii-kf18f70uf3, fmoc-leu, n-alpha-fmoc-l-leucine | |
| (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methylpentanoic acid |
Safety and Handling
TSCA : No