missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-leucine 98.0+%, TCI America™

Catalog No. p-7129595
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02951G 1 g
F02955G 5 g
F029525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F02951G Supplier TCI America Supplier No. F02951G
Only null left

Chemical Identifiers

CAS 35661-60-0
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD00037133
InChI Key CBPJQFCAFFNICX-TWYLJJHKNA-N
Synonym fmoc-l-leucine, fmoc-leu-oh, fmoc-l-leu-oh, n-fmoc-l-leucine, n-9-fluorenylmethoxycarbonyl-l-leucine, fmoc-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-l-leucine, unii-kf18f70uf3, fmoc-leu, n-alpha-fmoc-l-leucine
PubChem CID 1549133
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methylpentanoic acid
SMILES CC(C)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 154°C
Color White
Quantity 1 g
Formula Weight 353.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-leucine