missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine Hydrate 98.0+%, TCI America™

Supplier:  TCI America F030525G

 View more versions of this product

Catalog No. F030525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine Hydrate
White
MFCD00037139
fmoc-ala-oh, fmoc-l-alanine, fmoc-l-alpha-alanine, 9-fmoc-l-alanine, fmoc-l-ala-oh, n-fmoc-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-l-alanine, n-9-fluorenylmethoxycarbonyl-l-alanine, fmoc-alanine, fmoc-l-ala
CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
311.337
311.34
Crystalline Powder
35661-39-3
C18H17NO4
25 g
QWXZOFZKSQXPDC-NSHDSACASA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
6364642
≥98.0% (HPLC,T)

Chemical Identifiers

35661-39-3
311.337
QWXZOFZKSQXPDC-NSHDSACASA-N
6364642
CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C18H17NO4
MFCD00037139
fmoc-ala-oh, fmoc-l-alanine, fmoc-l-alpha-alanine, 9-fmoc-l-alanine, fmoc-l-ala-oh, n-fmoc-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-l-alanine, n-9-fluorenylmethoxycarbonyl-l-alanine, fmoc-alanine, fmoc-l-ala
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions