missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine Hydrate 98.0+%, TCI America™

Catalog No. p-7129603
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03055G 5 g
F030525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03055G Supplier TCI America Supplier No. F03055G
Only null left

Chemical Identifiers

CAS 35661-39-3
Molecular Formula C18H17NO4
Molecular Weight (g/mol) 311.337
MDL Number MFCD00037139
InChI Key QWXZOFZKSQXPDC-NSHDSACASA-N
Synonym fmoc-ala-oh, fmoc-l-alanine, fmoc-l-alpha-alanine, 9-fmoc-l-alanine, fmoc-l-ala-oh, n-fmoc-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-l-alanine, n-9-fluorenylmethoxycarbonyl-l-alanine, fmoc-alanine, fmoc-l-ala
PubChem CID 6364642
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Color White
Quantity 5 g
Formula Weight 311.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine Hydrate