missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]glycine 98.0+%, TCI America™
$42.87 - $76.43
Chemical Identifiers
| CAS | 29022-11-5 |
|---|---|
| Molecular Formula | C17H15NO4 |
| Molecular Weight (g/mol) | 297.31 |
| MDL Number | MFCD00037140 |
| InChI Key | NDKDFTQNXLHCGO-UHFFFAOYSA-N |
| Synonym | fmoc-gly-oh, fmoc-glycine, n-9h-fluoren-9-ylmethoxy carbonyl glycine, n-fmoc-glycine, fmoc-dl-glycine, n-9-fmoc-l-glycine, n-alpha-fmoc-glycine, fmoc-gly, n-9-fluorenylmethoxycarbonyl glycine |
| PubChem CID | 93124 |
| IUPAC Name | 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetic acid |
| SMILES | OC(=O)CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Chemical Identifiers
| 29022-11-5 | |
| 297.31 | |
| NDKDFTQNXLHCGO-UHFFFAOYSA-N | |
| 93124 | |
| OC(=O)CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| C17H15NO4 | |
| MFCD00037140 | |
| fmoc-gly-oh, fmoc-glycine, n-9h-fluoren-9-ylmethoxy carbonyl glycine, n-fmoc-glycine, fmoc-dl-glycine, n-9-fmoc-l-glycine, n-alpha-fmoc-glycine, fmoc-gly, n-9-fluorenylmethoxycarbonyl glycine | |
| 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetic acid |
Specifications
| 29022-11-5 | |
| White | |
| MFCD00037140 | |
| fmoc-gly-oh, fmoc-glycine, n-9h-fluoren-9-ylmethoxy carbonyl glycine, n-fmoc-glycine, fmoc-dl-glycine, n-9-fmoc-l-glycine, n-alpha-fmoc-glycine, fmoc-gly, n-9-fluorenylmethoxycarbonyl glycine | |
| OC(=O)CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 297.31 | |
| 297.31 | |
| Crystalline Powder |
| 176°C | |
| C17H15NO4 | |
| 5 g | |
| NDKDFTQNXLHCGO-UHFFFAOYSA-N | |
| 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetic acid | |
| 93124 | |
| ≥98.0% (HPLC,T) | |
| N-[(9H-Fluoren-9-ylmethoxy)carbonyl]glycine |
Safety and Handling
TSCA : No