missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-proline 98.0+%, TCI America™

Supplier:  TCI America F06065G

Catalog No. F06065G

Explore available promotions

Description

Description

Specifications

Specifications

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-proline
White-Yellow
MFCD00077067
fmoc-d-pro-oh, fmoc-d-proline, n-9h-fluoren-9-ylmethoxy carbonyl-d-proline, n-fmoc-d-proline, fmoc-d-pro, 2r-1-9h-fluoren-9-ylmethoxycarbonyl pyrrolidine-2-carboxylic acid, 2r-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, fmoc-3-pro-oh, ksc908s9r, n-alpha-fmoc-d-proline
C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O
337.375
337.38
Crystalline Powder
101555-62-8
C20H19NO4
5 g
ZPGDWQNBZYOZTI-GOSISDBHSA-N
(2R)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidine-2-carboxylic acid
688136
≥98.0% (HPLC)

Chemical Identifiers

101555-62-8
337.375
ZPGDWQNBZYOZTI-GOSISDBHSA-N
688136
C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O
C20H19NO4
MFCD00077067
fmoc-d-pro-oh, fmoc-d-proline, n-9h-fluoren-9-ylmethoxy carbonyl-d-proline, n-fmoc-d-proline, fmoc-d-pro, 2r-1-9h-fluoren-9-ylmethoxycarbonyl pyrrolidine-2-carboxylic acid, 2r-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, fmoc-3-pro-oh, ksc908s9r, n-alpha-fmoc-d-proline
(2R)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidine-2-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions