missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]anthranilic Acid 96.0+%, TCI America™

Catalog No. F09235G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F09235G Supplier TCI America Supplier No. F09235G

Chemical Identifiers

CAS 150256-42-1
Molecular Formula C22H17NO4
Molecular Weight (g/mol) 359.381
MDL Number MFCD00235883
InChI Key CNAVPEPPAVHHKN-UHFFFAOYSA-N
Synonym fmoc-2-abz-oh, fmoc-2-aminobenzoic acid, 2-fmoc-amino benzoic acid, fmoc-abz-oh, n-fmoc-anthranilic acid, 2-9h-fluoren-9-yl methoxy carbonyl amino benzoic acid, fmoc-anthranilic acid, benzoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino, 2-9h-fluoren-9-ylmethoxy carbonyl amino benzoic acid, n-2-9-fluorenylmethyloxycarbonyl amino-benzoic acid
PubChem CID 978386
IUPAC Name 2-(9H-fluoren-9-ylmethoxycarbonylamino)benzoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=CC=CC=C4C(=O)O

Specifications

Melting Point 213°C
Quantity 5 g
Formula Weight 359.38
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]anthranilic Acid