missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-aminobutyric Acid 98.0+%, TCI America™

$61.51 - $242.33

Chemical Identifiers

CAS 116821-47-7
Molecular Formula C19H19NO4
Molecular Weight (g/mol) 325.364
MDL Number MFCD00144889
InChI Key ACUIFAAXWDLLTR-UHFFFAOYSA-N
Synonym fmoc-gaba-oh, fmoc-gamma-abu-oh, fmoc-gaba, n-fmoc-dl-4-amino-butyric acid, 4-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-fmoc-4-aminobutyric acid, 4-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-gamma-fmoc-gamma-aminobutyric acid, 4-9h-fluoren-9-yl methoxy carbonylamino butanoic acid
PubChem CID 2756086
IUPAC Name 4-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
F09115G
View Documents
TCI America
F09115G
5 g N/A
Only null left
 
F091125G
View Documents
TCI America
F091125G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

116821-47-7
325.364
ACUIFAAXWDLLTR-UHFFFAOYSA-N
2756086
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCC(=O)O
C19H19NO4
MFCD00144889
fmoc-gaba-oh, fmoc-gamma-abu-oh, fmoc-gaba, n-fmoc-dl-4-amino-butyric acid, 4-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-fmoc-4-aminobutyric acid, 4-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-gamma-fmoc-gamma-aminobutyric acid, 4-9h-fluoren-9-yl methoxy carbonylamino butanoic acid
4-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid

Specifications

116821-47-7
White
MFCD00144889
fmoc-gaba-oh, fmoc-gamma-abu-oh, fmoc-gaba, n-fmoc-dl-4-amino-butyric acid, 4-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-fmoc-4-aminobutyric acid, 4-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-gamma-fmoc-gamma-aminobutyric acid, 4-9h-fluoren-9-yl methoxy carbonylamino butanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCC(=O)O
325.364
325.36
Crystalline Powder
174°C
C19H19NO4
5 g
ACUIFAAXWDLLTR-UHFFFAOYSA-N
4-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
2756086
≥98.0% (HPLC,T)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-aminobutyric Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'