accessibility menu, dialog, popup

UserName

OONHOOH

CAS RN 116821-47-7

IUPAC Name: 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms: 4-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol): 325.36
Molecular Formula: C19H19NO4
InChi Key: ACUIFAAXWDLLTR-UHFFFAOYSA-N
SMILES: OC(=O)CCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)