missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(6-Aminohexyl)-1-naphthalenesulfonamide Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 164°C |
| Color | White-Yellow |
| Quantity | 500 mg |
| Formula Weight | 342.88 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-(6-Aminohexyl)-1-naphthalenesulfonamide Hydrochloride |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |