accessibility menu, dialog, popup

UserName

HClH2NNHSOO

CAS RN 61714-25-8

IUPAC Name: hydrogen N-(6-aminohexyl)naphthalene-1-sulfonamide chloride
Synonyms: hydrogen N-(6-aminohexyl)naphthalene-1-sulfonamide chloride
Molecular Weight (g/mol): 342.88
Molecular Formula: C16H23ClN2O2S
InChi Key: HOCSVIGHWPLMFC-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCCCCCNS(=O)(=O)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)