missing translation for 'onlineSavingsMsg'
Learn More

N-(5-Aminopentyl)-2-nitrobenzenesulfonamide Hydrochloride 98.0+%, TCI America™

Catalog No. A16611G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A16611G Supplier TCI America Supplier No. A16611G
Only null left

Chemical Identifiers

CAS 437718-20-2
Molecular Formula C11H18ClN3O4S
Molecular Weight (g/mol) 323.792
MDL Number MFCD06797044
InChI Key USGKSPTYZQTIRH-UHFFFAOYSA-N
Synonym 1-Amino-5-(2-nitrobenzenesulfonamido)pentane Hydrochloride, N-(2-Nitrobenzenesulfonyl)-1,5-pentanediamine Hydrochloride, N-(o-Ns)-1,5-diaminopentane Hydrochloride
PubChem CID 11964112
IUPAC Name N-(5-aminopentyl)-2-nitrobenzenesulfonamide;hydrochloride
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)NCCCCCN.Cl

Specifications

Color Yellow
Quantity 1 g
Formula Weight 323.79
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(5-Aminopentyl)-2-nitrobenzenesulfonamide Hydrochloride