accessibility menu, dialog, popup

UserName

NOOSOOHNNH2HCl

CAS RN 437718-20-2

IUPAC Name: N-(5-aminopentyl)-2-nitrobenzene-1-sulfonamide hydrochloride
Synonyms: N-(5-aminopentyl)-2-nitrobenzenesulfonamide hydrochloride
Molecular Weight (g/mol): 323.79
Molecular Formula: C11H18ClN3O4S
InChi Key: USGKSPTYZQTIRH-UHFFFAOYSA-N
SMILES: Cl.NCCCCCNS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results