missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(4-Methoxybenzylidene)aniline 98.0+%, TCI America™
Supplier: TCI America M058225G
Specifications
| N-(4-Methoxybenzylidene)aniline | |
| 63°C | |
| MFCD00025836 | |
| n-4-methoxybenzylidene aniline, e-n-4-methoxybenzylidene aniline, n-p-anisal aniline, n-p-anisylideneaniline, 1-4-methoxyphenyl-n-phenylmethanimine, 4-methoxybenzyliden-anilin, 4-methoxybenzylideneaniline, 1e-1-4-methoxyphenyl-n-phenylmethanimine, p-methoxybenzylidene-phenyl-amine, n-4-methoxyphenyl methylidene aniline # | |
| COC1=CC=C(C=NC2=CC=CC=C2)C=C1 | |
| 211.26 | |
| 211.26 | |
| Crystalline Powder |
| 836-41-9 | |
| C14H13NO | |
| 25 g | |
| MSWPGMRTURVKRJ-UHFFFAOYSA-N | |
| 1-(4-methoxyphenyl)-N-phenylmethanimine | |
| 98633 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 836-41-9 | |
| 211.26 | |
| MSWPGMRTURVKRJ-UHFFFAOYSA-N | |
| 98633 | |
| COC1=CC=C(C=NC2=CC=CC=C2)C=C1 |
| C14H13NO | |
| MFCD00025836 | |
| n-4-methoxybenzylidene aniline, e-n-4-methoxybenzylidene aniline, n-p-anisal aniline, n-p-anisylideneaniline, 1-4-methoxyphenyl-n-phenylmethanimine, 4-methoxybenzyliden-anilin, 4-methoxybenzylideneaniline, 1e-1-4-methoxyphenyl-n-phenylmethanimine, p-methoxybenzylidene-phenyl-amine, n-4-methoxyphenyl methylidene aniline # | |
| 1-(4-methoxyphenyl)-N-phenylmethanimine |
Safety and Handling
TSCA : No