missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Methoxybenzylidene)aniline 98.0+%, TCI America™

Catalog No. p-7132250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M05825G 5 g
M058225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M05825G Supplier TCI America Supplier No. M05825G

Chemical Identifiers

CAS 836-41-9
Molecular Formula C14H13NO
Molecular Weight (g/mol) 211.26
MDL Number MFCD00025836
InChI Key MSWPGMRTURVKRJ-UHFFFAOYSA-N
Synonym n-4-methoxybenzylidene aniline, e-n-4-methoxybenzylidene aniline, n-p-anisal aniline, n-p-anisylideneaniline, 1-4-methoxyphenyl-n-phenylmethanimine, 4-methoxybenzyliden-anilin, 4-methoxybenzylideneaniline, 1e-1-4-methoxyphenyl-n-phenylmethanimine, p-methoxybenzylidene-phenyl-amine, n-4-methoxyphenyl methylidene aniline #
PubChem CID 98633
IUPAC Name 1-(4-methoxyphenyl)-N-phenylmethanimine
SMILES COC1=CC=C(C=NC2=CC=CC=C2)C=C1

Specifications

Melting Point 63°C
Quantity 5 g
Formula Weight 211.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Methoxybenzylidene)aniline