missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Fluorobenzyl)-4-methoxyaniline, 97%, Thermo Scientific™

$263.03 - $263.03

Chemical Identifiers

CAS 356531-43-6
Molecular Formula C14H14FNO
Molecular Weight (g/mol) 231.27
MDL Number MFCD03210757
InChI Key QHNUQKUZGHGTCA-UHFFFAOYSA-N
Synonym n-4-fluorobenzyl-4-methoxyaniline, n-4-fluorophenyl methyl-4-methoxyaniline, n-4-methoxyphenyl-4-fluorobenzenemethanamine
PubChem CID 964317
IUPAC Name N-[(4-fluorophenyl)methyl]-4-methoxyaniline
SMILES COC1=CC=C(C=C1)NCC2=CC=C(C=C2)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH56102MD
View Documents
Thermo Scientific Chemicals
H56102MD
250 mg
Each for $263.03
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
As pharmaceuticals, they find applications in antibiotics, as sedatives, and for cancer treatment. Ethers substituted with fluorine are employed as volatile anesthetics. In metered-dose inhalers, which are used to administer some asthma medications, flurorocarbons find application as propellants.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Chemical Identifiers

356531-43-6
231.27
QHNUQKUZGHGTCA-UHFFFAOYSA-N
964317
COC1=CC=C(C=C1)NCC2=CC=C(C=C2)F
C14H14FNO
MFCD03210757
n-4-fluorobenzyl-4-methoxyaniline, n-4-fluorophenyl methyl-4-methoxyaniline, n-4-methoxyphenyl-4-fluorobenzenemethanamine
N-[(4-fluorophenyl)methyl]-4-methoxyaniline

Specifications

356531-43-6
MFCD03210757
n-4-fluorobenzyl-4-methoxyaniline, n-4-fluorophenyl methyl-4-methoxyaniline, n-4-methoxyphenyl-4-fluorobenzenemethanamine
COC1=CC=C(C=C1)NCC2=CC=C(C=C2)F
231.27
231.26
N-(4-Fluorobenzyl)-4-methoxyaniline
C14H14FNO
250 mg
QHNUQKUZGHGTCA-UHFFFAOYSA-N
N-[(4-fluorophenyl)methyl]-4-methoxyaniline
964317
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents