accessibility menu, dialog, popup

UserName

CAS RN 356531-43-6

IUPAC Name: N-[(4-fluorophenyl)methyl]-4-methoxyaniline
Synonyms: N-[(4-fluorophenyl)methyl]-4-methoxyaniline
Molecular Weight (g/mol): 231.27
Molecular Formula: C14H14FNO
InChi Key: QHNUQKUZGHGTCA-UHFFFAOYSA-N
SMILES: COC1=CC=C(NCC2=CC=C(F)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results